Assay Details
| Gene Symbol: mldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258620.2 | NP_001245549.1 | 1 - 2 | 1368 | 65 | |
| NM_132081.4 | NP_572309.2 | 1 - 2 | 1368 | 65 | ||
| GenBank mRNA | BT150142.1 | - | 1 - 2 | 1314 | 65 | |
More Information
Set Membership: |
|

mldr
Genomic Map