Assay Details
| Gene Symbol: CG34417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103423.3 | NP_001096893.1 | 6 - 7 | 9187 | 64 | |
| NM_001103425.3 | NP_001096895.1 | 6 - 7 | 9187 | 64 | ||
| NM_001103426.3 | NP_001096896.1 | 6 - 7 | 9187 | 64 | ||
| NM_001169208.3 | NP_001162679.1 | 6 - 7 | 9187 | 64 | ||
| NM_001169209.3 | NP_001162680.1 | 6 - 7 | 9187 | 64 | ||
| NM_001201619.3 | NP_001188548.1 | 5 - 6 | 8916 | 64 | ||
| NM_001201620.2 | NP_001188549.1 | 5 - 6 | 8836 | 64 | ||
| NM_001272347.1 | NP_001259276.1 | 5 - 6 | 4407 | 64 | ||
| NM_001272348.1 | NP_001259277.1 | 4 - 5 | 4056 | 64 | ||
| NM_001272350.1 | NP_001259279.1 | 6 - 7 | 9187 | 64 | ||
| NM_001272351.1 | NP_001259280.1 | 6 - 7 | 9187 | 64 | ||
| NM_001272352.1 | NP_001259281.1 | 6 - 7 | 9187 | 64 | ||
| NM_001272353.1 | NP_001259282.1 | 6 - 7 | 9187 | 64 | ||
| NM_001272354.1 | NP_001259283.1 | 6 - 7 | 9187 | 64 | ||
| NM_001272355.1 | NP_001259284.1 | 6 - 7 | 9187 | 64 | ||
| NM_001298017.1 | NP_001284946.1 | 4 - 5 | 8565 | 64 | ||
| NM_001298018.1 | NP_001284947.1 | 5 - 6 | 8916 | 64 | ||
| NM_001298019.1 | NP_001284948.1 | 4 - 5 | 8565 | 64 | ||
More Information
Set Membership: |
|

CG34417
Genomic Map