Assay Details
| Gene Symbol: Pat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272356.2 | NP_001259285.1 | 4 - 5 | 1366 | 102 | |
| NM_001272357.2 | NP_001259286.1 | 4 - 5 | 1366 | 102 | ||
| NM_132100.5 | NP_572328.1 | 4 - 5 | 1366 | 102 | ||
| GenBank mRNA | AY058334.1 | - | 4 - 5 | 1192 | 102 | |
More Information
Set Membership: |
|

Pat1
Genomic Map