Assay Details
| Gene Symbol: Fum1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132111.3 | NP_572339.1 | 1 - 2 | 194 | 54 | |
| GenBank mRNA | BT126329.1 | - | 1 - 2 | 161 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fum1| Gene Symbol: Fum1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132111.3 | NP_572339.1 | 1 - 2 | 194 | 54 | |
| GenBank mRNA | BT126329.1 | - | 1 - 2 | 161 | 54 | |
Set Membership: |
|