Assay Details
| Gene Symbol: C3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144694.2 | NP_001138166.1 | 7 - 8 | 1900 | 67 | |
| NM_001144696.3 | NP_001138168.1 | 6 - 7 | 2307 | 67 | ||
| NM_001272362.1 | NP_001259291.1 | 6 - 7 | 2151 | 67 | ||
| NM_132122.5 | NP_572350.2 | 6 - 7 | 2970 | 67 | ||
More Information
Set Membership: |
|

C3G
Genomic Map