Assay Details
| Gene Symbol: iav | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132125.2 | NP_572353.1 | 2 - 3 | 1195 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

iav| Gene Symbol: iav | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132125.2 | NP_572353.1 | 2 - 3 | 1195 | 79 | |
Set Membership: |
|