Assay Details
| Gene Symbol: unc-119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298049.1 | NP_001284978.1 | 4 - 5 | 1440 | 76 | |
| NM_132161.5 | NP_572389.1 | 4 - 5 | 1444 | 76 | ||
| GenBank mRNA | AY118580.1 | - | 4 - 5 | 1423 | 76 | |
More Information
Set Membership: |
|

unc-119
Genomic Map