Assay Details
| Gene Symbol: NELF-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132174.3 | NP_572402.1 | 1 - 2 | 209 | 59 | |
| GenBank mRNA | AF145659.1 | - | 1 - 2 | 171 | 59 | |
More Information
Set Membership: |
|

NELF-B
Genomic Map