Assay Details
| Gene Symbol: Nek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132187.3 | NP_572415.1 | 1 | 2416 | 159 | |
| GenBank mRNA | AY095023.1 | - | 1 | 2346 | 159 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nek2| Gene Symbol: Nek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132187.3 | NP_572415.1 | 1 | 2416 | 159 | |
| GenBank mRNA | AY095023.1 | - | 1 | 2346 | 159 | |
Set Membership: |
|