Assay Details
| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272388.1 | NP_001259317.1 | 3 - 4 | 2679 | 109 | |
| NM_001272389.1 | NP_001259318.1 | 4 - 5 | 2619 | 109 | ||
| NM_132200.4 | NP_572428.3 | 3 - 4 | 2679 | 109 | ||
More Information
Set Membership: |
|

Ubr3
Genomic Map