Assay Details
| Gene Symbol: l(1)G0020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298096.1 | NP_001285025.1 | 3 - 4 | 2044 | 74 | |
| NM_132275.2 | NP_572503.1 | 3 - 4 | 2044 | 74 | ||
| GenBank mRNA | BT132962.1 | - | 3 - 4 | 2026 | 74 | |
More Information
Set Membership: |
|

l(1)G0020
Genomic Map