Assay Details
| Gene Symbol: Usp15-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138115.3 | NP_611959.2 | 1 - 2 | 1092 | 56 | |
| GenBank mRNA | BT057999.1 | - | 1 - 2 | 1029 | 56 | |
More Information
Set Membership: |
|

Usp15-31
Genomic Map