Assay Details
| Gene Symbol: Usp15-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299926.1 | NP_001286855.1 | 3 - 4 | 1596 | 81 | |
| NM_001299927.1 | NP_001286856.1 | 3 - 4 | 1596 | 81 | ||
| NM_138115.3 | NP_611959.2 | 4 - 5 | 1680 | 81 | ||
| GenBank mRNA | BT057999.1 | - | 4 - 5 | 1617 | 81 | |
More Information
Set Membership: |
|

Usp15-31
Genomic Map