Assay Details
| Gene Symbol: CG6845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138156.5 | NP_612000.3 | 4 - 5 | 1065 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG6845| Gene Symbol: CG6845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138156.5 | NP_612000.3 | 4 - 5 | 1065 | 68 | |
Set Membership: |
|