Assay Details
| Gene Symbol: CG7028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274257.1 | NP_001261186.1 | 2 - 3 | 575 | 100 | |
| NM_001299955.1 | NP_001286884.1 | 2 - 3 | 575 | 100 | ||
| NM_001299956.1 | NP_001286885.1 | 2 - 3 | 575 | 100 | ||
| NM_001299957.1 | NP_001286886.1 | 2 - 3 | 575 | 100 | ||
| NM_138166.4 | NP_612010.1 | 2 - 3 | 575 | 100 | ||
| GenBank mRNA | AF145626.1 | - | 2 - 3 | 548 | 100 | |
More Information
Set Membership: |
|

CG7028
Genomic Map