Assay Details
| Gene Symbol: Cdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169842.2 | NP_001163313.1 | 2 - 3 | 445 | 58 | |
| NM_138189.3 | NP_612033.1 | 2 - 3 | 572 | 58 | ||
| GenBank mRNA | AY061302.1 | - | 2 - 3 | 392 | 58 | |
More Information
Set Membership: |
|

Cdc5
Genomic Map