Assay Details
| Gene Symbol: Trh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138236.2 | NP_612080.1 | 3 - 4 | 813 | 72 | |
| GenBank mRNA | BT001418.1 | - | 3 - 4 | 789 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trh| Gene Symbol: Trh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138236.2 | NP_612080.1 | 3 - 4 | 813 | 72 | |
| GenBank mRNA | BT001418.1 | - | 3 - 4 | 789 | 72 | |
Set Membership: |
|