Assay Details
| Gene Symbol: otk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138561.3 | NP_612571.1 | 1 - 2 | 464 | 77 | |
| GenBank mRNA | AY089628.1 | - | 1 - 2 | 462 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

otk2| Gene Symbol: otk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138561.3 | NP_612571.1 | 1 - 2 | 464 | 77 | |
| GenBank mRNA | AY089628.1 | - | 1 - 2 | 462 | 77 | |
Set Membership: |
|