Assay Details
| Gene Symbol: dsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104007.4 | NP_001097477.1 | 6 - 7 | 1644 | 86 | |
| NM_139418.3 | NP_647675.3 | 5 - 6 | 1180 | 86 | ||
| GenBank mRNA | BT058102.1 | - | 6 - 7 | 1644 | 86 | |
More Information
Set Membership: |
|

dsb
Genomic Map