Assay Details
| Gene Symbol: Dbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139420.4 | NP_647677.2 | 2 - 3 | 1334 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbx| Gene Symbol: Dbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139420.4 | NP_647677.2 | 2 - 3 | 1334 | 91 | |
Set Membership: |
|