Assay Details
| Gene Symbol: Mfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139422.3 | NP_647679.1 | 2 - 3 | 1112 | 56 | |
| GenBank mRNA | AY047562.1 | - | 2 - 3 | 1118 | 56 | |
More Information
Set Membership: |
|

Mfap1
Genomic Map