Assay Details
| Gene Symbol: ProRS-m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274408.1 | NP_001261337.1 | 3 - 4 | 722 | 62 | |
| NM_139480.3 | NP_647737.1 | 3 - 4 | 714 | 62 | ||
| GenBank mRNA | BT150200.1 | - | 3 - 4 | 651 | 62 | |
More Information
Set Membership: |
|

ProRS-m
Genomic Map