Assay Details
| Gene Symbol: Shab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202108.2 | NP_001189037.1 | 11 - 12 | 5077 | 119 | |
| NM_001202109.2 | NP_001189038.1 | 10 - 11 | 4867 | 119 | ||
| NM_001202110.2 | NP_001189039.1 | 10 - 11 | 4942 | 119 | ||
| NM_001274416.1 | NP_001261345.1 | 10 - 11 | 4972 | 119 | ||
More Information
Set Membership: |
|

Shab
Genomic Map