Assay Details
| Gene Symbol: Shab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202108.2 | NP_001189037.1 | 12 - 13 | 5347 | 101 | |
| NM_001202110.2 | NP_001189039.1 | 11 - 12 | 5212 | 101 | ||
| NM_001274416.1 | NP_001261345.1 | 11 - 12 | 5242 | 101 | ||
More Information
Set Membership: |
|

Shab
Genomic Map