Assay Details
| Gene Symbol: ND-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139518.3 | NP_647775.1 | 1 - 2 | 172 | 73 | |
| GenBank mRNA | AY118532.1 | - | 1 - 2 | 133 | 73 | |
| BT011342.1 | - | 1 - 2 | 168 | 73 | ||
More Information
Set Membership: |
|

ND-30
Genomic Map