Assay Details
| Gene Symbol: CG14965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139526.4 | NP_647783.3 | 1 | 1760 | 152 | |
| GenBank mRNA | AY052024.1 | - | 1 | 1726 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG14965| Gene Symbol: CG14965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139526.4 | NP_647783.3 | 1 | 1760 | 152 | |
| GenBank mRNA | AY052024.1 | - | 1 | 1726 | 152 | |
Set Membership: |
|