Assay Details
| Gene Symbol: akirin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202129.2 | NP_001189058.1 | 2 - 3 | 575 | 69 | |
| NM_001202130.2 | NP_001189059.1 | 2 - 3 | 706 | 69 | ||
| NM_001202131.2 | NP_001189060.1 | 2 - 3 | 565 | 69 | ||
| NM_001274582.1 | NP_001261511.1 | 2 - 3 | 590 | 69 | ||
| NM_139856.4 | NP_648113.1 | 2 - 3 | 706 | 69 | ||
| NM_168211.3 | NP_729249.1 | 2 - 3 | 590 | 69 | ||
| GenBank mRNA | AY095189.1 | - | 2 - 3 | 689 | 69 | |
More Information
Set Membership: |
|

akirin
Genomic Map