Assay Details
| Gene Symbol: Pdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274616.1 | NP_001261545.1 | 2 - 3 | 2519 | 71 | |
| NM_001274618.1 | NP_001261547.1 | 3 - 4 | 324 | 71 | ||
| NM_001300044.1 | NP_001286973.1 | 3 - 4 | 1430 | 71 | ||
| NM_080093.5 | NP_524832.3 | 3 - 4 | 1430 | 71 | ||
| NM_168233.3 | NP_729297.1 | 2 - 3 | 282 | 71 | ||
| NM_168234.3 | NP_729298.2 | 3 - 4 | 1430 | 71 | ||
| NM_168235.3 | NP_729299.1 | 2 - 3 | 2519 | 71 | ||
| NM_168236.3 | NP_729300.1 | 3 - 4 | 2561 | 71 | ||
| NM_168237.2 | NP_729301.1 | 4 - 5 | 2156 | 71 | ||
| NM_168238.3 | NP_729302.2 | 3 - 4 | 2097 | 71 | ||
| NM_168239.2 | NP_729303.1 | 3 - 4 | 2200 | 71 | ||
| GenBank mRNA | AY070526.1 | - | 4 - 5 | 2156 | 71 | |
| BT003290.1 | - | 2 - 3 | 343 | 71 | ||
More Information
Set Membership: |
|

Pdp1
Genomic Map