Assay Details
| Gene Symbol: pbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168242.3 | NP_729307.1 | 1 - 2 | 390 | 65 | |
| NM_168243.3 | NP_729308.1 | 1 - 2 | 390 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pbl| Gene Symbol: pbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168242.3 | NP_729307.1 | 1 - 2 | 390 | 65 | |
| NM_168243.3 | NP_729308.1 | 1 - 2 | 390 | 65 | ||
Set Membership: |
|