Assay Details
| Gene Symbol: ImpE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274648.1 | NP_001261577.1 | 3 - 4 | 679 | 100 | |
| NM_080110.3 | NP_524849.2 | 3 - 4 | 623 | 100 | ||
| NM_168270.2 | NP_729371.1 | 3 - 4 | 623 | 100 | ||
| GenBank mRNA | AY095083.1 | - | 3 - 4 | 619 | 100 | |
More Information
Set Membership: |
|

ImpE1
Genomic Map