Assay Details
| Gene Symbol: Zasp66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168283.4 | NP_729395.2 | 6 - 7 | 1122 | 67 | |
| NM_168284.3 | NP_729396.2 | 6 - 7 | 1111 | 67 | ||
| NM_168286.4 | NP_729398.1 | 5 - 6 | 609 | 67 | ||
| NM_176306.3 | NP_788484.2 | 4 - 5 | 749 | 67 | ||
| GenBank mRNA | AY058415.1 | - | 6 - 7 | 1089 | 67 | |
| BT088849.1 | - | 5 - 6 | 585 | 67 | ||
More Information
Set Membership: |
|

Zasp66
Genomic Map