Assay Details
| Gene Symbol: Zasp66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169911.1 | NP_001163382.1 | 6 - 7 | 1195 | 85 | |
| NM_001169914.3 | NP_001163385.1 | 6 - 7 | 1206 | 85 | ||
| NM_168283.4 | NP_729395.2 | 7 - 8 | 1278 | 85 | ||
| NM_168284.3 | NP_729396.2 | 7 - 8 | 1267 | 85 | ||
| NM_168286.4 | NP_729398.1 | 6 - 7 | 765 | 85 | ||
| NM_176303.5 | NP_788481.1 | 5 - 6 | 693 | 85 | ||
| NM_176306.3 | NP_788484.2 | 5 - 6 | 905 | 85 | ||
| GenBank mRNA | AY058415.1 | - | 7 - 8 | 1245 | 85 | |
| BT029127.1 | - | 6 - 7 | 1175 | 85 | ||
| BT088849.1 | - | 6 - 7 | 741 | 85 | ||
| BT133470.1 | - | 6 - 7 | 1204 | 85 | ||
More Information
Set Membership: |
|

Zasp66
Genomic Map