Assay Details
| Gene Symbol: Zasp66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169911.1 | NP_001163382.1 | 1 - 2 | 300 | 94 | |
| NM_001169913.1 | NP_001163384.1 | 1 - 2 | 300 | 94 | ||
| NM_139989.4 | NP_648246.3 | 1 - 2 | 300 | 94 | ||
| NM_168284.3 | NP_729396.2 | 1 - 2 | 300 | 94 | ||
| GenBank mRNA | AY058415.1 | - | 1 - 2 | 278 | 94 | |
| BT029127.1 | - | 1 - 2 | 280 | 94 | ||
More Information
Set Membership: |
|

Zasp66
Genomic Map