Assay Details
| Gene Symbol: Zasp66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168286.4 | NP_729398.1 | 2 - 3 | 257 | 86 | |
| NM_176303.5 | NP_788481.1 | 2 - 3 | 257 | 86 | ||
| NM_176304.5 | NP_788482.1 | 2 - 3 | 257 | 86 | ||
| NM_176305.4 | NP_788483.1 | 2 - 3 | 257 | 86 | ||
| NM_176306.3 | NP_788484.2 | 1 - 2 | 397 | 86 | ||
| GenBank mRNA | BT088849.1 | - | 2 - 3 | 233 | 86 | |
| BT089012.1 | - | 2 - 3 | 252 | 86 | ||
More Information
Set Membership: |
|

Zasp66
Genomic Map