Assay Details
| Gene Symbol: Cpr66D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168288.3 | NP_729400.1 | 1 - 2 | 401 | 122 | |
| GenBank mRNA | AY071527.1 | - | 1 - 2 | 366 | 122 | |
More Information
Set Membership: |
|

Cpr66D
Genomic Map