Assay Details
| Gene Symbol: Prm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168291.1 | NP_729406.1 | 3 - 4 | 1270 | 67 | |
| NM_168292.2 | NP_729407.1 | 3 - 4 | 1270 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prm| Gene Symbol: Prm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168291.1 | NP_729406.1 | 3 - 4 | 1270 | 67 | |
| NM_168292.2 | NP_729407.1 | 3 - 4 | 1270 | 67 | ||
Set Membership: |
|