Assay Details
| Gene Symbol: eIF4E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274697.1 | NP_001261626.1 | 3 - 4 | 524 | 83 | |
| NM_001274698.1 | NP_001261627.1 | 2 - 3 | 825 | 83 | ||
| NM_080090.3 | NP_524829.1 | 2 - 3 | 516 | 83 | ||
| NM_168334.1 | NP_729481.1 | 2 - 3 | 462 | 83 | ||
| NM_168335.2 | NP_729482.1 | 2 - 3 | 755 | 83 | ||
| NM_168336.2 | NP_729483.1 | 2 - 3 | 556 | 83 | ||
| NM_168337.2 | NP_729484.1 | 2 - 3 | 484 | 83 | ||
| NM_168338.2 | NP_729485.1 | 2 - 3 | 390 | 83 | ||
| GenBank mRNA | BT044219.1 | - | 2 - 3 | 730 | 83 | |
| U16139.1 | - | 1 - 2 | 235 | 83 | ||
More Information
Set Membership: |
|

eIF4E1
Genomic Map