Assay Details
| Gene Symbol: S-Lap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168411.2 | NP_729613.1 | 1 | 1954 | 148 | |
| GenBank mRNA | AY061822.1 | - | 1 | 1887 | 148 | |
| BT003650.1 | - | 3413 | 148 | |||
More Information
Set Membership: |
|

S-Lap4
Genomic Map