Assay Details
| Gene Symbol: Alg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168449.2 | NP_729680.1 | 1 - 2 | 256 | 68 | |
| GenBank mRNA | AJ431376.1 | - | 1 - 2 | 251 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg10| Gene Symbol: Alg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168449.2 | NP_729680.1 | 1 - 2 | 256 | 68 | |
| GenBank mRNA | AJ431376.1 | - | 1 - 2 | 251 | 68 | |
Set Membership: |
|