Assay Details
| Gene Symbol: btl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014583.2 | NP_001014583.1 | 2 - 3 | 2795 | 62 | |
| NM_168577.3 | NP_729956.1 | 3 - 4 | 2947 | 62 | ||
More Information
Set Membership: |
|

btl
Genomic Map