Assay Details
| Gene Symbol: GlyRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274938.1 | NP_001261867.1 | 2 | 2341 | 85 | |
| NM_140489.3 | NP_648746.1 | 2 | 2322 | 85 | ||
| NM_168605.2 | NP_730022.1 | 1 | 2410 | 85 | ||
| GenBank mRNA | AY051413.1 | - | 2 | 2308 | 85 | |
| BT133207.1 | - | 1 | 2370 | 85 | ||
More Information
Set Membership: |
|

GlyRS
Genomic Map