Assay Details
| Gene Symbol: brm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274977.1 | NP_001261906.1 | 3 - 4 | 459 | 59 | |
| NM_080497.5 | NP_536745.4 | 3 - 4 | 447 | 59 | ||
| NM_080498.3 | NP_536746.1 | 3 - 4 | 459 | 59 | ||
| NM_168640.2 | NP_730088.1 | 3 - 4 | 442 | 59 | ||
| NM_168641.1 | NP_730089.1 | 2 - 3 | 673 | 59 | ||
| GenBank mRNA | AY095048.1 | - | 3 - 4 | 457 | 59 | |
| BT009972.1 | - | 3 - 4 | 447 | 59 | ||
More Information
Set Membership: |
|

brm
Genomic Map