Assay Details
| Gene Symbol: dsx-c73A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168667.3 | NP_730169.1 | 1 - 2 | 1818 | 86 | |
| GenBank mRNA | AY071185.1 | - | 1 - 2 | 1814 | 86 | |
More Information
Set Membership: |
|

dsx-c73A
Genomic Map