Assay Details
| Gene Symbol: Shark | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080004.4 | NP_524743.2 | 2 - 3 | 628 | 83 | |
| GenBank mRNA | AY051937.1 | - | 2 - 3 | 603 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shark| Gene Symbol: Shark | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080004.4 | NP_524743.2 | 2 - 3 | 628 | 83 | |
| GenBank mRNA | AY051937.1 | - | 2 - 3 | 603 | 83 | |
Set Membership: |
|