Assay Details
| Gene Symbol: Shark | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080004.4 | NP_524743.2 | 3 - 4 | 1155 | 77 | |
| GenBank mRNA | AY051937.1 | - | 3 - 4 | 1130 | 77 | |
More Information
Set Membership: |
|

Shark
Genomic Map