Assay Details
| Gene Symbol: RFeSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272947.1 | NP_001259876.1 | 1 - 2 | 174 | 79 | |
| NM_080009.4 | NP_524748.2 | 1 - 2 | 174 | 79 | ||
| NM_164426.3 | NP_722715.1 | 1 - 2 | 174 | 79 | ||
| GenBank mRNA | BT122041.1 | - | 1 - 2 | 153 | 79 | |
More Information
Set Membership: |
|

RFeSP
Genomic Map