Assay Details
| Gene Symbol: GluRIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274692.1 | NP_001261621.2 | 6 - 7 | 1558 | 84 | |
| NM_001274693.1 | NP_001261622.1 | 6 - 7 | 1558 | 84 | ||
| NM_001316480.1 | NP_001303409.1 | 6 - 7 | 1558 | 84 | ||
| NM_080016.4 | NP_524755.2 | 6 - 7 | 1558 | 84 | ||
| GenBank mRNA | BT150463.1 | - | 6 - 7 | 1557 | 84 | |
More Information
Set Membership: |
|

GluRIB
Genomic Map