Assay Details
| Gene Symbol: Tollo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080018.2 | NP_524757.1 | 1 | 7073 | 140 | |
| GenBank mRNA | AY118551.1 | - | 1 | 7049 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tollo| Gene Symbol: Tollo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080018.2 | NP_524757.1 | 1 | 7073 | 140 | |
| GenBank mRNA | AY118551.1 | - | 1 | 7049 | 140 | |
Set Membership: |
|