Assay Details
| Gene Symbol: RpL37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201762.1 | NP_001188691.1 | 3 - 4 | 260 | 59 | |
| NM_080042.4 | NP_524781.1 | 3 - 4 | 217 | 59 | ||
| NM_164628.3 | NP_723060.1 | 2 - 3 | 289 | 59 | ||
| GenBank mRNA | AY071189.1 | - | 3 - 4 | 213 | 59 | |
| AY071725.1 | - | 3 - 4 | 214 | 59 | ||
| KX532150.1 | - | 1 - 2 | 134 | 59 | ||
More Information
Set Membership: |
|

RpL37A
Genomic Map