Assay Details
| Gene Symbol: Bap111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132302.4 | NP_572530.1 | 1 | 2537 | 140 | |
| GenBank mRNA | AJ002520.1 | - | 1 | 2500 | 140 | |
| BT016087.1 | - | 1 | 2521 | 140 | ||
More Information
Set Membership: |
|

Bap111
Genomic Map